Orders LinkContactUsLinkComponent
Bench mounted gas taps
Bench mounted gas taps
2× propane/butane (LPG), EPDM seal
Catalog # KOTT733-00056
Supplier:   KOETTERMANN
Bench mounted gas taps
Catalog # KOTT733-00056
Supplier:  KOETTERMANN
Bench mounted gas taps
2× propane/butane (LPG), EPDM seal
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
These bench mounted gas taps are sealed by EPDM (ethylene propylene diene terpolymer) which is an extremely durable synthetic rubber roofing membrane.
Specifications
  • Description:
    2× propane/butane (LPG), EPDM seal
  • Seq:
    02