Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Color Disc Replacement, Iron LR (FerroVer), Outdoor-Light, pk/1 0 … 1.0 mg/l 1 * 1 items
Catalog # HACH9263900
Supplier:
Hach
Expand image
Color Disc Replacement, Iron LR (FerroVer), Outdoor-Light, pk/1 0 … 1.0 mg/l 1 * 1 items
Catalog # HACH9263900
Supplier:
Hach