Volumetric flasks, class A, white markings
15 ml, 10/19, 0,04 ± ml
WITG3.670.016
Borosilicate glass 3.3, amber, with glass stopper.
- White markings
- Conformity certified (DE-M marking)
- Inclusive batch certificate
: DIN EN ISO 1042
- NS:10/19
- Capacity:15 ml
- Seq:04
- Tolerance:0,04 ± ml