Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DULBECCO'S MODIFICATION OF EAGLE'S MEDIUM 1 * 100 mL
Catalog # ICNA091642149
Supplier:
MP Biomedicals
Expand image
DULBECCO'S MODIFICATION OF EAGLE'S MEDIUM 1 * 100 mL
Catalog # ICNA091642149
Supplier:
MP Biomedicals