Order Entry
Export
Orders LinkContactUsLinkComponent
 

α-Cyclodextrin, white powder

25 g
Catalog # ICNA0219005225
Supplier:   MP Biomedicals
α-Cyclodextrin, white powder
Catalog # ICNA0219005225
Supplier:  MP Biomedicals
CAS Number:  10016-20-3
α-Cyclodextrin, white powder

25 g

CAS Number:  10016-20-3
Price
361,00 €
Log in for availability
1 * 25 g
Product Details & Documents

α-Cyclodextrin is reported useful for the selective precipitation of enantiomeric, positional or structural isomers. Cyclodextrins have hydrophobic cavities; form inclusion compounds with organic substances, salts and halogens in the solid state or in aqueous solutions.

Soluble in water (50 mg/ml).

  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Melting Pt:278 °C (lit.)
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Loss on Drying:
    ≤13%
  • Optical Rotation:
    +148 ± 5°(c=1, water)

Specifications

  • Pk:
    25 g

Specifications