Order Entry
Export
Orders LinkContactUsLinkComponent
 

L-thyroxine, powder

5 g
Catalog # ICNA0215214505
Supplier:   MP Biomedicals
L-thyroxine, powder
Catalog # ICNA0215214505
Supplier:  MP Biomedicals
CAS Number:  51-48-9
L-thyroxine, powder

5 g

CAS Number:  51-48-9
Price
470,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Soluble in solutions of alkali hydroxides (50 mg/ml in 4 M ammonium hydroxide in methanol); slightly insoluble in alcohol; insoluble in water, acetone, chloroform and ether.

Synonyms:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS Number:51-48-9
  • Formula:C₁₅H₁₁I₄NO₄
  • MDL Number:MFCD00002595
  • Molecular Weight:776.874 g/mol
Specifications

Specifications

  • Pk:
    5 g

Specifications