L-thyroxine, powder
500 mg
Catalog # ICNA02152145.5
Supplier: MP Biomedicals
Jump to:
Product Details & Documents
Soluble in solutions of alkali hydroxides (50 mg/ml in 4 M ammonium hydroxide in methanol); slightly insoluble in alcohol; insoluble in water, acetone, chloroform and ether.
Synonyms: (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
- CAS Number:51-48-9
- Formula:C₁₅H₁₁I₄NO₄
- MDL Number:MFCD00002595
- Molecular Weight:776.874 g/mol
Specifications
Specifications
- Pk:500 mg



