Oligomycin
5 mg
Catalog # ICNA0215178605
Supplier: MP Biomedicals
Jump to:
Product Details & Documents
Soluble in ether, benzene, absolute ethanol, glacial acetic acid and acetone.
Synonyms: Oligomycin A, B, C, Oligomycin complex
- CAS Number:1404-19-9
- Formula:C₄₅H₇₄O₁₁
- MDL Number:MFCD01779388
- Molecular Weight:789.3 g/mol
Specifications
Specifications
- Pk:5 mg



