ContactUsLinkComponent
MP SILICA, 18-32 µM, 60Å. Mean Pore Size: 60ÅSpecific Surface Area: 500-600 m2 /gParticle Size: 18-32 µmWeight: 0.4-0.5 g/mlpH: 7.0 1 * 500 g
Catalog # ICNA0202753
Supplier:  MP Biomedicals
MP SILICA, 18-32 µM, 60Å. Mean Pore Size: 60ÅSpecific Surface Area: 500-600 m2 /gParticle Size: 18-32 µmWeight: 0.4-0.5 g/mlpH: 7.0 1 * 500 g
Catalog # ICNA0202753
Supplier:  MP Biomedicals