Order Entry
Export
Orders LinkContactUsLinkComponent
Hydroxyurea, crystalline powder

Hydroxyurea, crystalline powder

Glass bottle for solids, 10 g
Catalog # ICNA0210202310
Supplier:   MP Biomedicals
Hydroxyurea, crystalline powder
Catalog # ICNA0210202310
Supplier:  MP Biomedicals
CAS Number:  127-07-1
Hydroxyurea, crystalline powder

Glass bottle for solids, 10 g

CAS Number:  127-07-1
Price
198,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Hydroxyurea is a inhibitor of DNA synthesis in vivo, and an antiviral and antineoplastic agent which is S-phase specific.

A ribonucleotide reductase inhibitor - inhibits by forming a free radical nitroxide that binds a tyrosyl free radical in the active site of the enzyme. Induces fetal hemoglobin production in sickle cell anemia patients.

It induces apoptosis of in vitro myelogenous leukemia lymphocytes, and the phosphorylation of FADD and p53. It is also used to reduce the rate of painful attacks in sickle-cell disease and has antiretroviral properties in diseases such as AIDS.

It is Anti-neoplastic. It inactivates ribonucleoside reductase by forming a free radical nitroxide that binds a tyrosyl free radical in the active site of the enzyme. This blocks the synthesis of deoxynucleotides, which inhibits DNA synthesis and induces synchronisation or cell death in S-phase.

Freely soluble in water (50 mg/ml-clear colourless solution) or hot alcohol.

Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:13,04874
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:136…146 °C
  • Storage Temperature:Ambient
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications