ContactUsLinkComponent
Purity: 99% Crystalline, Used in treatment of Parkinsonism. Also an inhibitor of tyrosine aminotransferase. 1 * 250 mg
Catalog # ICNA02101578.2
Supplier:  MP Biomedicals
Purity: 99% Crystalline, Used in treatment of Parkinsonism. Also an inhibitor of tyrosine aminotransferase. 1 * 250 mg
Catalog # ICNA02101578.2
Supplier:  MP Biomedicals