Order Entry
Export
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) ≥99%, powder

3,3',5,5'-Tetramethylbenzidine (TMB) ≥99%, powder

1 g
Catalog # ICNA0219027701
Irritant
Supplier:   MP Biomedicals
3,3',5,5'-Tetramethylbenzidine (TMB) ≥99%, powder
Catalog # ICNA0219027701
Supplier:  MP Biomedicals
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) ≥99%, powder

1 g

CAS Number:  54827-17-7
Price
71,00 €
Log in for availability
1 * 1 g
Product Details & Documents

3,3',5,5'-Tetramethylbenzidine is suitable as peroxidase substrate for use in ELISA procedures. The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620 to 650 nm. The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm. A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases. It is useful in gel staining procedure for low levels of home-associated peroxidases and for enzyme immunoassay of horseradish peroxidase.

Soluble in solvents 5%, Ethyl Acetate; insoluble in water. Dissolve 1,5 mg TMB in 0,1 ml of DMSO. Then add 9,9 ml of 0,1 M sodium acetate (pH 6,0). Finally, add hydrogen peroxide to a concentration of 0,01% and filter through a Whatman #1 paper filter. Prepare solutions fresh daily. Discard any remaining solution at the end of the day.

Caution: Research Use Only (RUO).

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • Melting Point:
    165 - 170 °C

Specifications

  • Pk:
    1 g

Specifications