Order Entry
Export
Orders LinkContactUsLinkComponent
 

Gentamicin sulphate

1 g
Catalog # ICNA0219453001
Health
Supplier:   MP Biomedicals
Gentamicin sulphate
Catalog # ICNA0219453001
Supplier:  MP Biomedicals
CAS Number:  1405-41-0
Gentamicin sulphate

1 g

CAS Number:  1405-41-0
Price
213,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Gentamicin is an aminoglycoside antibiotic complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is a mixture of 3 major components designated as C1, C1a, and C2. Gentamicin is used as the sulfate salt. Each component consists of five basic nitrogens and requires five equivalents of sulfuric acid per mole of gentamicin base. Mode of action: Gentamicin causes codon misreading by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site.The bactericidal effect of gentamicin on Pseudomonas aeruginosa is exerted by the binding of gentamicin to the outer membrane, where it displaces natural cations, destabilizes the membrane, and forms holes in the cell surface.Antimicrobial spectrum: Gram-negative bacteria, Staphylococcus aureus and other Gram-positive bacteria. Gentamicin sulfate is a broad spectrum antibiotic. It inhibits the growth of a wide variety of Gram-positive and Gram-negative microorganisms, including strains resistant to tetracycline, chloramphenicol, kanamycin, and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus, and Streptococcus. Gentamicin sulfate inhibits bacterial protein biosynthesis by binding to the 30S subunit of the ribosome.

  • Gentamicin sulfate is soluble in water (50 mg/mL), yielding a clear to very slightly hazy, colorless to faint. It is practically insoluble in alcohol and other organic solvents.

Storage temperature: +4 °C.

Synonyms:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • MDL Number:MFCD00270181
  • Merck Index:13,04403
  • Melting Pt:218…237 °C (lit.)
  • Storage Temperature:Ambient
  • EINECS:215-778-9
Specifications

Specifications

  • Pk:
    1 g

Specifications