Order Entry
Export
Orders LinkContactUsLinkComponent
Dimethyl sulphoxide ≥99%, liquid

Dimethyl sulphoxide ≥99%, liquid

100 mL
Catalog # ICNA0219018680
Supplier:   MP Biomedicals
Dimethyl sulphoxide ≥99%, liquid
Catalog # ICNA0219018680
Supplier:  MP Biomedicals
CAS Number:  67-68-5
Dimethyl sulphoxide ≥99%, liquid
Dimethyl sulphoxide ≥99%, liquid

100 mL

CAS Number:  67-68-5
Price
29,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Dimethyl sulfoxide is a dipolar, aprotic solvent. Has been shown to accelerate strand renaturation (1-10% concentration) and is believed to give the nucleic acid thermal stability against depurination. By heating the solid form to approximately 30 °C, the product can be melted without harming it's stability.

  • Auto Ignition: 573 °F
  • Vapor Pressure: 0.42 mmHg ( 20 °C) (Lit.)
  • Refractive Index: n20/D 1.479(lit.)
  • Dielectric Constant: 45

Dimethyl sulfoxide is used to enhance dermal absorption of many chemicals. A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers.Used in antifreeze or hydraulic fluids and as a cryopreservative for cell cultures.Used in the oxidation of thiols and disulfides to sulfonic acids. Used as a PCR cosolvent to help improve yields, especially in long PCR.

Soluble in water, methanol, acetone, ether, benzene, chloroform.

Caution: Store at Room Temperature (15-30 °C), desiccate.

Synonyms:  Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
  • CAS Number:67-68-5
  • Formula:(CH₃)₂SO
  • MDL Number:MFCD00002089
  • Molecular Weight:78.14 g/mol
  • Boiling Pt:189 °C (1013 hPa) (lit.)
  • Melting Pt:16…19 °C (lit.)
  • Density:1.104 g/cm³ (20 °C) (lit.)
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    >99%

Specifications

  • Pk:
    100 mL

Specifications