Order Entry
Export
Orders LinkContactUsLinkComponent
Bathophenanthroline, white, powder

Bathophenanthroline, white, powder

Glass bottle for solids, 1 g
Catalog # ICNA0219015701
Supplier:   MP Biomedicals
Bathophenanthroline, white, powder
Catalog # ICNA0219015701
Supplier:  MP Biomedicals
CAS Number:  1662-01-7
Bathophenanthroline, white, powder

Glass bottle for solids, 1 g

CAS Number:  1662-01-7
Price
200,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Bathophenanthroline is used as a bidentate ligand and for the colorimetric determination of iron.

Bathophenanthroline is used as a bidentate ligand and for the colorimetric determination of iron.

Soluble in 95% ethanol:chloroform (2:1) (50 mg/ml. Insoluble in water.

Synonyms:  4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
  • CAS Number:1662-01-7
  • Formula:C₂₄H₁₆N₂
  • MDL Number:MFCD00004976
  • Molecular Weight:332.4 g/mol
  • Boiling Pt:508 °C (1013 hPa)
  • Melting Pt:215…224 °C
  • Storage Temperature:Ambient
  • EINECS:216-767-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications