Accessories for refrigerated circulating bath VersaCool™
Stainless steel, adjustable VersaRack, holds up to 75 test tubes of 10 mm diameter, 48 test tubes of 16 mm diameter and 21 test tubes of 25 mm diameter
Catalog # HAKA230000001
Supplier: THERMO ELECTRON LED
Accessories for refrigerated circulating bath VersaCool™
Catalog # HAKA230000001
Supplier: THERMO ELECTRON LED
Jump to:
Product Details & Documents
Specifications
- Description:Stainless steel, adjustable VersaRack, holds up to 75 test tubes of 10 mm diameter, 48 test tubes of 16 mm diameter and 21 test tubes of 25 mm diameter
- Seq:07