ContactUsLinkComponent
pH combination puncture electrode, gel-filled, pH 2…14, 0 … 60°C open diaphragm, POM/Glass body, S7 Top connector 1 * 1 items
Catalog # HACHLZW5233970002
Supplier:  Hach
pH combination puncture electrode, gel-filled, pH 2…14, 0 … 60°C open diaphragm, POM/Glass body, S7 Top connector 1 * 1 items
Catalog # HACHLZW5233970002
Supplier:  Hach
Related Information