ContactUsLinkComponent
pH combination electrode, gel-filled, pH 0…14, 0 … 60°C PTFE sleeve diaphragm, Glass body, S7 Top connector 1 * 1 items
Catalog # HACHLZW5223970002
Supplier:  Hach
pH combination electrode, gel-filled, pH 0…14, 0 … 60°C PTFE sleeve diaphragm, Glass body, S7 Top connector 1 * 1 items
Catalog # HACHLZW5223970002
Supplier:  Hach