ContactUsLinkComponent
Pipettes graduated,VOLAC,USP,1ml:0.01,AS,BS ISO 835, cap.at outlet,tot.del.,indiv.calibrated, works certificate 1 * 5 items
Catalog # POULUS385/WAC/A/5
Supplier:  Poulten & Graf
undefined
Pipettes graduated,VOLAC,USP,1ml:0.01,AS,BS ISO 835, cap.at outlet,tot.del.,indiv.calibrated, works certificate 1 * 5 items
Catalog # POULUS385/WAC/A/5
Supplier:  Poulten & Graf