ContactUsLinkComponent
pH combination electrode, gel-filled, pH 0…14, 0…80°C 1x ceramic diaphragm, Polycarbonate/Glass body body, 1 m cable, BNC connector 1 * 1 items
Catalog # HACHLZW5010970002
Supplier:  Hach
pH combination electrode, gel-filled, pH 0…14, 0…80°C 1x ceramic diaphragm, Polycarbonate/Glass body body, 1 m cable, BNC connector 1 * 1 items
Catalog # HACHLZW5010970002
Supplier:  Hach