Order Entry
Export
Orders LinkContactUsLinkComponent
12% TRIMETHYLAMINESOLUTION 1 * 200 mL

12% TRIMETHYLAMINESOLUTION 1 * 200 mL

Catalog # WAKO200-20021
Supplier:   Wako
12% TRIMETHYLAMINESOLUTION 1 * 200 mL
Catalog # WAKO200-20021
Supplier:  Wako
CAS Number:  
12% TRIMETHYLAMINESOLUTION 1 * 200 mL
CAS Number:  
Price
406,00 €
Log in for availability
1 * 200 mL
Product Details & Documents