Electronic comparators, EComparator series
EC 3000 Saybolt
Catalog # TINT163020
Supplier: Lovibond Tintometer
Jump to:
Product Details & Documents
The Lovibond® EComparator series provides an easy way to transition from subjective visual measurement to a non subjective, accurate electronic measurement.
- Digital, portable, push-button technology
- On-screen colour
- On-screen numerical display
- User friendly ergonomics and intuitive interface guarantee new users can be quickly trained and easily supported
- Large data storage (>20000 readings)
- USB connectivity ensures readings can be stored and shared easily and quickly
Delivery information: Instrument supplied with USB cable, USB power supply, 4× AA batteries, 2× liquid reference solutions, glass reference standards, cell divider, glass and plastic cells, mini screwdriver all contained in a plastic carrying case.
Specifications
- Description:EC 3000 Saybolt
- Cat. No.:TINT163020
- Light source:White LED
- Repeatability:±1
- Parameters:Saybolt
- Standard conformances:ASTM D156, D6045
- Model:EC 3000 Saybolt
- W×D×H:106×210×57 mm
- Power supply:USB or 4×AA batteries
- Range:−16...+30
- Path length:50 mm
- Comparator view:3 fields
- Resolution:1 Saybolt unit



