Order Entry
Export
Orders LinkContactUsLinkComponent
Electronic comparators, EComparator series

Electronic comparators, EComparator series

EC 2000 Gardner
Catalog # TINT162020
Supplier:   Lovibond Tintometer
Electronic comparators, EComparator series
Catalog # TINT162020
Electronic comparators, EComparator series
Electronic comparators, EComparator series

EC 2000 Gardner

Price
3 510,00 €
Log in for availability
1 * 1 items
Product Details & Documents

The Lovibond® EComparator series provides an easy way to transition from subjective visual measurement to a non subjective, accurate electronic measurement.

  • Digital, portable, push-button technology
  • On-screen colour
  • On-screen numerical display
  • User friendly ergonomics and intuitive interface guarantee new users can be quickly trained and easily supported
  • Large data storage (>20000 readings)
  • USB connectivity ensures readings can be stored and shared easily and quickly

Delivery information: Instrument supplied with USB cable, USB power supply, 4× AA batteries, 2× liquid reference solutions, glass reference standards, cell divider, glass and plastic cells, mini screwdriver all contained in a plastic carrying case.

Specifications
  • Description:
    EC 2000 Gardner
  • Cat. No.:
    TINT162020
  • Light source:
    White LED
  • Repeatability:
    ±0,2
  • Parameters:
    Gardner colour scale
  • Standard conformances:
    ASTM D1544, D6166
  • Model:
    EC 2000 Gardner
  • W×D×H:
    106×210×57 mm
  • Power supply:
    USB or 4×AA batteries
  • Range:
    0 - 18
  • Path length:
    10 mm
  • Comparator view:
    2 fields
  • Resolution:
    0,1 Gardner unit