0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Microscope Objectives
Objectives for CKX53
Objectives for CKX53
Objectives for CKX53
Dry objective with pre-centre phase contrast slider, 2,2 mm
OLYMN5229400
:
OLYMPUS OPTICAL
Objectives for CKX53
OLYMN5229400
:
OLYMPUS OPTICAL
Expand image
Objectives for CKX53
Dry objective with pre-centre phase contrast slider, 2,2 mm
Add to cart
724,00 €
Unit of Measure
1 * 1 items
-
+
Add to cart
Magnification:
Ph 40×/0,55
Description:
Dry objective with pre-centre phase contrast slider
Seq:
023
Working distance:
2,2 mm