Orders LinkContactUsLinkComponent
KP-moulds
KP-moulds
Moulds, 15×15×5 mm
Catalog # KLIN3051
Supplier:   KLINIPATH
KP-moulds
Catalog # KLIN3051
Supplier:  KLINIPATH
KP-moulds
Moulds, 15×15×5 mm
Price
73,00 €
Log in for availability
1 * 12 items
Product Details & Documents
These moulds are made of stainless steel.
Available in various sizes.
Specifications
  • Description:
    Moulds
  • Dimensions:
    15×15×5 mm