Order Entry
Export
Orders LinkContactUsLinkComponent
Tank liners, Corning®

Tank liners, Corning®

200 L, LDPE, 57,15×91,44 mm
Catalog # MDTC91-300-30
Supplier:   Corning
Tank liners, Corning®
Catalog # MDTC91-300-30
Supplier:  Corning
Tank liners, Corning®

200 L, LDPE, 57,15×91,44 mm

Price
128,47 €
Log in for availability
1 * 1 items
Product Details & Documents

Corning® tank liners are designed to fit cylindrical tanks.

  • Available in a range of sizes

Tank liners are manufactured from LDPE.

Specifications
  • Ø×H:
    57,15×91,44 mm
  • Capacity:
    200 L
  • Seq:
    04
  • Tubing material:
    LDPE