Orders LinkContactUsLinkComponent
VWR®, Dropping Bottles
VWR®, Dropping Bottles
Pipette kit for 30 ml dropping bottles, -
Catalog # 215-3826
Supplier:   VWR Collection
VWR®, Dropping Bottles
Catalog # 215-3826
Supplier:  VWR Collection
VWR®, Dropping Bottles
VWR®, Dropping Bottles
Pipette kit for 30 ml dropping bottles, -
Price
71,10 €
Log in for availability
1 * 100 items
Product Details & Documents
Soda-lime glass, amber.
  • Good chemical and physical properties
  • Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.
Specifications
  • Capacity:
    -
  • Description:
    Pipette kit for 30 ml dropping bottles
  • Seq:
    11