Precision laboratory thermometer stem form Capillaries form: yellow enamelled round liquid filling: spec.red range: 0+50C in 0.1C Total length: 420 mm immersion length: total, suitable for DAkkS certification 1 * 1 items
Catalog # SCNL1042354/04/3P
Supplier: Ludwig Schneider GmbH & CO. KG