Order Entry
Export
Orders LinkContactUsLinkComponent
 

Gentamicin sulphate

Bottle, 10 g
Catalog # APOSBIG0124-10G
Irritant
Health
Supplier:   Apollo Scientific
Gentamicin sulphate
Catalog # APOSBIG0124-10G
Supplier:  Apollo Scientific
CAS Number:  1405-41-0
Gentamicin sulphate

Bottle, 10 g

CAS Number:  1405-41-0
Price
24,60 €
Log in for availability
1 * 10 g
Product Details & Documents

An aminoglycoside antibiotic.

  • approx. 650 units/mg
  • Cell culture tested
Synonyms:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • MDL Number:MFCD00270181
  • Melting Pt:217…237 °C
  • Storage Temperature:Refrigerator
  • EINECS:215-778-9

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications