Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
An agonist of human CAR (EC50 = 49 nM) 1 * 10 mg
Catalog # CAYM16027-10
Supplier:
Cayman Chemical
Expand image
An agonist of human CAR (EC50 = 49 nM) 1 * 10 mg
Catalog # CAYM16027-10
Supplier:
Cayman Chemical