Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
BDP-12 1 * 10 mg
BDP-12 1 * 10 mg
Print
BDP-12 1 * 10 mg
Catalog # CAYM14936-10
Supplier:
Cayman Chemical
BDP-12 1 * 10 mg
Catalog # CAYM14936-10
Supplier:
Cayman Chemical
CAS Number:
Expand image
Return to Top
BDP-12 1 * 10 mg
Add to cart
CAS Number:
Price
135,00 €
Log in
for availability
Unit of Measure
1 * 10 mg
-
+
Add to cart
Return Policy
Product Details & Documents