Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
An orally bioavailable inhibitor of MEK1/2 (IC50 = 12 nM) that inhibits ERK phosphorylation in various cancer cell lines (IC50s 5 nM) 1 * 5 mg
CAYM16996-5
:
Cayman Chemical
An orally bioavailable inhibitor of MEK1/2 (IC50 = 12 nM) that inhibits ERK phosphorylation in various cancer cell lines (IC50s 5 nM) 1 * 5 mg
CAYM16996-5
:
Cayman Chemical