ContactUsLinkComponent
An ATP-competitive Pim-1 kinase inhibitor (IC50 = 50 nM) that displays selectivity over the related kinases, Pim-2 and MEK1/2 (IC50s = >20 µM) 1 * 5 mg
Catalog # CAYM16286-5
Supplier:  Cayman Chemical
An ATP-competitive Pim-1 kinase inhibitor (IC50 = 50 nM) that displays selectivity over the related kinases, Pim-2 and MEK1/2 (IC50s = >20 µM) 1 * 5 mg
Catalog # CAYM16286-5
Supplier:  Cayman Chemical