Order Entry
Export
Orders LinkContactUsLinkComponent
GC guard columns, HydroGOLD

GC guard columns, HydroGOLD

Guard column, 0,32 mm, 10 m
Catalog # 555-9116
GC guard columns, HydroGOLD
Catalog # 555-9116
GC guard columns, HydroGOLD

Guard column, 0,32 mm, 10 m

Price
220,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These guard columns extend analytical column lifetime by preventing degradation from harsh 'steam cleaning' water injections.

  • Highly deactivated to provide superior inertness, essential for analysis of active compounds
  • Protects against column contamination caused by non-volatile materials
Specifications
  • Column Format:
    Guard column
  • Ø int.:
    0,32 mm
  • Length:
    10 m