VWR®, Microtube Storage Boxes
100, Natural, 141×151×47 mm
Catalog # 525-0926
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP.
- Highly transparent, with bold identifying colours and a distinctive wave design
- Uniform wells with location ID; capacity indicated on the outside of the box
- Black light-tight version available for light-sensitive samples
- Frosted labelling patch
- Alphanumeric identification on the bottom
Compact footprint for microtube and cryogenic vial storage. Store 1,5/2,0 ml microtubes or cryogenic vials. Complete with an easy open, friction-fit lid. Will fit into most standard freezer racking.
Delivery information: Assorted pack contains one of each colour blue, green, pink, yellow and orange.
Specifications
- Compartments:100
- W×D×H ext.:141×151×47 mm
- Colour:Natural
- Seq:12