Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Electrochemistry Meters
pH and Multi-Parameter Meters
Accessories for Lovibond® Multi Meter, Handheld, SD 40
Accessories for Lovibond® Multi Meter, Handheld, SD 40
Print
Accessories for Lovibond® Multi Meter, Handheld, SD 40
pH buffer set
Catalog # LOVI721250
Supplier:
Lovibond
Accessories for Lovibond® Multi Meter, Handheld, SD 40
Catalog # LOVI721250
Supplier:
Lovibond
Expand image
Return to Top
Accessories for Lovibond® Multi Meter, Handheld, SD 40
pH buffer set
Specifications
Add to cart
Jump to:
Specifications
Price
18,20 €
Log in
for availability
Unit of Measure
1 * 1 SET
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Specifications
Description:
pH buffer set
Seq:
03