Calcium acetate USP
Poly pail, 12 kg
SPCMC1067-12KG
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
: Calcium acetate, United States Pharmacopeia (USP) Reference Standard, Calcium di(acetate), Acetic acid calcium salt
- :62-54-4
- :(H₃CCOO)₂Ca
- :MFCD00150019
- :158.17 g/mol
- :12,01683
- :1.509 g/cm³ (20 °C)
- :Ambient
- :200-540-9
- Assay:99% Min.
- Pack type:Poly pail
- Pk:12 kg



