Order Entry
Export
Orders LinkContactUsLinkComponent
L(+)-Valine USP

L(+)-Valine USP

Poly bottle, 1 kg
Catalog # SPCMV1005-1KG
Supplier:   Spectrum Chemical
L(+)-Valine USP
Catalog # SPCMV1005-1KG
Supplier:  Spectrum Chemical
CAS Number:  72-18-4
L(+)-Valine USP

Poly bottle, 1 kg

CAS Number:  72-18-4
Price
840,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  H-Val-OH, Valine, (S)-(+)-Valine
  • CAS Number:72-18-4
  • Formula:C₅H₁₁NO₂
  • MDL Number:MFCD00064220
  • Molecular Weight:117.15 g/mol
  • Merck Index:13,09458
  • Boiling Pt:217 °C (1013 hPa)
  • Melting Pt:86 °C
  • Density:1.284 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-773-6

Documents

Specifications

Specification Test Results

  • Assay:
    98.5% Min.

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    1 kg

Specifications