1-Octadecanol NF
Fiber drum, 45 kg
Catalog # SPCMST115-45KGBL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
- CAS Number:112-92-5
- Formula:H₃C(CH₂)₁₆CH₂OH
- MDL Number:MFCD00002823
- Molecular Weight:270.5 g/mol
- Merck Index:12,08960
- Boiling Pt:336 °C (1013 hPa)
- Melting Pt:56…57 °C
- Density:0.805…0.815 g/cm³ (60 °C)
- Storage Temperature:Ambient
- EINECS:204-017-6
Specifications
Specification Test Results
- Assay:90% Min.
Specifications
- Pack type:Fiber drum
- Pk:45 kg



