(±)-1,3-Butanediol
Steel pail, 20 L
Catalog # SPCMB2147-20LT
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: Butane-1,3-diol, (±)-1,3-Butylene glycol, (±)-1,3-Dihydroxy butane
- CAS Number:107-88-0
- Formula:C₄H₁₀O₂
- MDL Number:MFCD00004554
- Molecular Weight:90.12 g/mol
- Merck Index:13,01566
- Boiling Pt:206…208 °C (1013 hPa)
- Melting Pt:< –50 °C
- Density:1.004…1.006 g/cm³ (20 °C)
- Flash Pt:116 °C
- Storage Temperature:Ambient
- EINECS:203-529-7
Specifications
Specification Test Results
- Assay:99% Min.
Specifications
- Pack type:Steel pail
- Pk:20 L



