Sorbitan monooleate NF
Glass, amber, 4 L
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: Sorbitan oleate, Span 80, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol mono-(9Z)-9-octadecenoate
- CAS Number:1338-43-8
- Formula:C₂₄H₄₄O₆
- MDL Number:MFCD00080948
- Molecular Weight:428.61 g/mol
- Storage Temperature:Ambient
- EINECS:215-665-4
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Assay:72% Min.
Specifications
- Pack type:Glass, amber
- Pk:4 L



