Polysorbate 60 NF
200 L
Catalog # SPCMPO136-200LTBL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: Sorbitan monostearate, ethoxylated, Polyoxyethylene sorbitan monostearate
- CAS Number:9005-67-8
- Formula:C₆₄H₁₂₆O₂₆
- MDL Number:MFCD01780110
- Storage Temperature:Ambient
- EINECS:500-020-4
Specifications
Specifications
- Pk:200 L



