Agar, powder NF
Poly bottle, 2,5 kg
Catalog # SPCMAG110-2.5KG
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Form: Powder
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- Storage Temperature:Ambient
- EINECS:232-658-1
Specifications
Specifications
- Pack type:Poly bottle
- Pk:2,5 kg



