Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Butyraldehyde 1 * 200 L
Print
Butyraldehyde 1 * 200 L
Catalog # SPCMB2271-200LTBL
Supplier:
Spectrum Chemical
Butyraldehyde 1 * 200 L
Catalog # SPCMB2271-200LTBL
Supplier:
Spectrum Chemical
CAS Number:
Return to Top
Butyraldehyde 1 * 200 L
CAS Number:
Price
6 750,00 €
Log in
for availability
1 * 200 L
-
+
Return Policy
Product Details & Documents