Order Entry
Export
Orders LinkContactUsLinkComponent
Dextrin, yellow, powder

Dextrin, yellow, powder

50 lb
Catalog # SPCMDE138-50LBBL
Supplier:   Spectrum Chemical
Dextrin, yellow, powder
Catalog # SPCMDE138-50LBBL
Supplier:  Spectrum Chemical
CAS Number:  9004-53-9
Dextrin, yellow, powder

50 lb

CAS Number:  9004-53-9
Price
1 180,00 €
Log in for availability
1 * 50 lb
Product Details & Documents

Form: Yellow Powder

Synonyms:  Dextrin
  • CAS Number:9004-53-9
  • Formula:(C₆H₁₂O₆)n
  • MDL Number:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Storage Temperature:Ambient
  • EINECS:232-675-4
Specifications

Specifications

  • Pk:
    50 lb

Specifications