Dextrin, yellow, powder
Poly bottle, 2,5 kg
Catalog # SPCMDE138-2.5KG
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Form: Yellow Powder
Synonyms: Dextrin
- CAS Number:9004-53-9
- Formula:(C₆H₁₂O₆)n
- MDL Number:MFCD00081554
- Molecular Weight:180.16 g/mol
- Storage Temperature:Ambient
- EINECS:232-675-4
Specifications
Specifications
- Pack type:Poly bottle
- Pk:2,5 kg
- Cat. No.:SPCMDE138-2.5KG



