Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl phthalate NF

Diethyl phthalate NF

Glass, amber, 4 L
Catalog # SPCMD1097-4LTGL
Supplier:   Spectrum Chemical
Diethyl phthalate NF
Catalog # SPCMD1097-4LTGL
Supplier:  Spectrum Chemical
CAS Number:  84-66-2
Diethyl phthalate NF
Diethyl phthalate NF

Glass, amber, 4 L

CAS Number:  84-66-2
Price
515,00 €
Log in for availability
1 * 4 L
Product Details & Documents

Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Synonyms:  Phthalic acid diethyl ester
  • CAS Number:84-66-2
  • Formula:C₁₂H₁₄O₄
  • MDL Number:MFCD00009111
  • Molecular Weight:222.24 g/mol
  • Merck Index:12,07527
  • Boiling Pt:298 °C (1013 hPa)
  • Melting Pt:–3 °C
  • Density:1.12 g/cm³ (20 °C)
  • Flash Pt:156 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:201-550-6

Documents

Specifications

Specification Test Results

  • Assay:
    98% Min.

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    4 L

Specifications