Diethyl phthalate NF
200 L
Catalog # SPCMD1097-200LTBL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: Phthalic acid diethyl ester
- CAS Number:84-66-2
- Formula:C₁₂H₁₄O₄
- MDL Number:MFCD00009111
- Molecular Weight:222.24 g/mol
- Merck Index:12,07527
- Boiling Pt:298 °C (1013 hPa)
- Melting Pt:–3 °C
- Density:1.12 g/cm³ (20 °C)
- Flash Pt:156 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:201-550-6
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Assay:98% Min.
Specifications
- Pk:200 L
- Cat. No.:SPCMD1097-200LTBL



