Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SONOREX DIGIPLUS DL 1028 H, ultrasonic bath with power settings, 35 kHz, 1 * 1 items
Catalog # BAND7186-CH
Supplier:
BANDELIN-ELECTRONIC
Expand image
SONOREX DIGIPLUS DL 1028 H, ultrasonic bath with power settings, 35 kHz, 1 * 1 items
Catalog # BAND7186-CH
Supplier:
BANDELIN-ELECTRONIC