Order Entry
Export
Orders LinkContactUsLinkComponent
Dispersing tools for T 65

Dispersing tools for T 65

D 60, -/60 mm, 2 - 30 l, 520 mm
Catalog # IKAA20008388
Supplier:   IKA
Dispersing tools for T 65
Catalog # IKAA20008388
Supplier:  IKA
Dispersing tools for T 65

D 60, -/60 mm, 2 - 30 l, 520 mm

Price
627,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Volume:
    2 - 30 l
  • Shaft length:
    520 mm
  • Stator/rotor Ø:
    -/60 mm
  • Type:
    D 60